Skip to Content
MilliporeSigma
Cymiazole PESTANAL®, analytical standard

Cymiazole

Cymiazole PESTANAL®, analytical standard
Synonym(s):
2,4-Dimethyl-N-(3-methyl-2(3H)-thiazolylidene)benzenamine, 2-(2,4-Dimethylphenylimino)-3-methylthiazoline, 2-(2,4-Xylylimino)-3-methylthiazoline, N-[(2EZ)-3-Methyl-1,3-thiazol-2-(3H)ylidene]-2,4-xylidine
Empirical Formula (Hill Notation):
C12H14N2S
CAS Number:
Molecular Weight:
218.32
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing