Skip to Content
MilliporeSigma
Cyhalothrin analytical standard

Cyhalothrin

Cyhalothrin analytical standard
Empirical Formula (Hill Notation):
C23H19ClF3NO3
CAS Number:
Molecular Weight:
449.85
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing