Skip to Content
MilliporeSigma
Cyclen tetrahydrochloride 98%

Cyclen tetrahydrochloride

Cyclen tetrahydrochloride 98%
Synonym(s):
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
Empirical Formula (Hill Notation):
C8H20N4 · 4HCl
CAS Number:
Molecular Weight:
318.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing