Skip to Content
MilliporeSigma
Cryolite synthetic, ≥97.0% (from F)

Cryolite

Cryolite synthetic, ≥97.0% (from F)
Synonym(s):
Kryolith, Sodium hexafluoroaluminate
Linear Formula:
Na3AlF6
CAS Number:
Molecular Weight:
209.94

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing