Skip to Content
MilliporeSigma
Crotonaldehyde, predominantly trans ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer

Crotonaldehyde, predominantly trans

Crotonaldehyde, predominantly trans ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Linear Formula:
CH3CH=CHCHO
CAS Number:
Molecular Weight:
70.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1