Skip to Content
MilliporeSigma
Coumarin 6 ≥99%

Coumarin 6

Coumarin 6 ≥99%
Synonym(s):
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin, 3-(2-Benzothiazolyl)-N,N-diethylumbelliferylamine
Empirical Formula (Hill Notation):
C20H18N2O2S
CAS Number:
Molecular Weight:
350.43
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing