Skip to Content
MilliporeSigma
Coumarin 102 Dye content 98 %

Coumarin 102

Coumarin 102 Dye content 98 %
Synonym(s):
2,3,6,7-Tetrahydro-9-methyl-1H,5H-quinolizino(9,1-gh)coumarin, 8-Methyl-2,3,5,6-tetrahydro-1H,4H-11-oxa-3a-aza-benzo(de)anthracen-10-one, Coumarin 480
Empirical Formula (Hill Notation):
C16H17NO2
CAS Number:
Molecular Weight:
255.31
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing