Skip to Content
MilliporeSigma
Coproporphyrin I tetramethyl ester ≥90% (HPLC)

Coproporphyrin I tetramethyl ester

Coproporphyrin I tetramethyl ester ≥90% (HPLC)
Synonym(s):
Tetramethyl 3,8,13,18-tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionate
Empirical Formula (Hill Notation):
C40H46N4O8
CAS Number:
Molecular Weight:
710.82
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing