Skip to Content
MilliporeSigma
Copper(II) nitrate hemi(pentahydrate) ACS reagent, ≥99.99% trace metals basis

Copper(II) nitrate hemi(pentahydrate)

Copper(II) nitrate hemi(pentahydrate) ACS reagent, ≥99.99% trace metals basis
Synonym(s):
Cupric nitrate hemi(pentahydrate)
Linear Formula:
Cu(NO3)2 · 2.5H2O
CAS Number:
Molecular Weight:
232.59
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade