Skip to Content
MilliporeSigma
Copper Chlorophyllin Pharmaceutical Secondary Standard; Certified Reference Material

Copper Chlorophyllin

Copper Chlorophyllin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Chlorophyllin sodium copper salt, Chlorophyllin, coppered trisodium salt
Empirical Formula (Hill Notation):
C34H31CuN4Na3O6
CAS Number:
Molecular Weight:
724.15
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1