Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Conessine
Synonym(s):
(3β)-N,N-dimethyl-con-5-enin-3-amine, Conessin, NSC 119994, Neriine, Roquessine, Wrightine
Empirical Formula (Hill Notation):
C
24
H
40
N
2
CAS Number:
546-06-5
Molecular Weight:
356.59
EC Number:
208-897-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
SML0095
Conessine,
≥97% (HPLC)
Expand
View Pricing