Skip to Content
MilliporeSigma
Coenzyme B12 ≥97.0%

Coenzyme B12

Coenzyme B12 ≥97.0%
Synonym(s):
5′-Deoxyadenosylcobalamine, Adenosyl cobalamine, Cobamamide, DMBC coenzyme
Empirical Formula (Hill Notation):
C72H100CoN18O17P
CAS Number:
Molecular Weight:
1579.58
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing