Skip to Content
MilliporeSigma
Coenzyme A, oxidized lithium salt ≥85%

Coenzyme A, oxidized lithium salt

Coenzyme A, oxidized lithium salt ≥85%
Synonym(s):
CoA-S-S-CoA
Empirical Formula (Hill Notation):
C42H70N14O32P6S2 · xLi+
CAS Number:
Molecular Weight:
1533.05 (free acid basis)

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing