Skip to Content
MilliporeSigma
Cocaethylene solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Cocaethylene solution

Cocaethylene solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C18H23NO4
CAS Number:
Molecular Weight:
317.38

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique