Skip to Content
MilliporeSigma
Cobalt(III) acetylacetonate 99.99% trace metals basis

Cobalt(III) acetylacetonate

Cobalt(III) acetylacetonate 99.99% trace metals basis
Synonym(s):
Co(acac)3, Cobalt(III) 2,4-pentanedionate, Cobaltic acetylacetonate, Tris(2,4-pentanedionato)cobalt, Cobalt(III) acetylacetonate
Linear Formula:
Co(C5H7O2)3
CAS Number:
Molecular Weight:
356.26

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Core
Showing 1-3 of 3