Skip to Content
MilliporeSigma
Clofarabine ≥98% (HPLC)

Clofarabine

Clofarabine ≥98% (HPLC)
Synonym(s):
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol, 2-chloro-2′-arabino-fluoro-2′-deoxyadenosine, (2-Chloro-9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl)-9H-purin-6-amine)
Empirical Formula (Hill Notation):
C10H11ClFN5O3
CAS Number:
Molecular Weight:
303.68

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing