Skip to Content
MilliporeSigma
Clenbuterol Related Compound B certified reference material, pharmaceutical secondary standard

Clenbuterol Related Compound B

Clenbuterol Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanone hydrochloride
Empirical Formula (Hill Notation):
C12H16Cl2N2O · xHCl
CAS Number:
Molecular Weight:
275.17 (free base basis)

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade