
Clarithromycin Impurity K

Synonym(s):
(1S,2R,5R,6S,7S,8R,9R,Z)-8-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-6-hydroxy-9-methoxy-1,5,7,9,11,13-hexamethyl-3,15-dioxabicyclo[10.2.1]pentadeca-11,13-dien-4-one
Empirical Formula (Hill Notation):
C30H51NO8
CAS Number:
Molecular Weight:
553.73
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing