Skip to Content
MilliporeSigma
CITCO ≥98% (HPLC), solid

CITCO

CITCO ≥98% (HPLC), solid
Synonym(s):
6-(4-Chlorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde O-(3,4-dichlorobenzyl)oxime
Empirical Formula (Hill Notation):
C19H12N3OCl3S
CAS Number:
Molecular Weight:
436.74

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing