Skip to Content
MilliporeSigma
cis-Diamineplatinum(II) dichloride ≥99.9% trace metals basis

cis-Diamineplatinum(II) dichloride

cis-Diamineplatinum(II) dichloride ≥99.9% trace metals basis
Synonym(s):
cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Activity Spectrum
Quality Segment
Markush Class
Markush Group
Mode of Action
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing