Skip to Content
MilliporeSigma
cis-4-Benzyloxy-2-buten-1-ol ≥95%

cis-4-Benzyloxy-2-buten-1-ol

cis-4-Benzyloxy-2-buten-1-ol ≥95%
Synonym(s):
cis-2-Buten-1,4-diol-monobenzyl ether
Linear Formula:
C6H5CH2OCH2CH=CHCH2OH
CAS Number:
Molecular Weight:
178.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing