Skip to Content
MilliporeSigma
cis-1-Bromo-2-ethoxyethylene technical, ~85% (GC)

cis-1-Bromo-2-ethoxyethylene

cis-1-Bromo-2-ethoxyethylene technical, ~85% (GC)
Synonym(s):
cis-2-Ethoxyvinyl bromide
Empirical Formula (Hill Notation):
C4H7BrO
CAS Number:
Molecular Weight:
151.00
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing