Skip to Content
MilliporeSigma
cis-11,14,17-Eicosatrienoic acid methyl ester ≥98%, liquid

cis-11,14,17-Eicosatrienoic acid methyl ester

cis-11,14,17-Eicosatrienoic acid methyl ester ≥98%, liquid
Empirical Formula (Hill Notation):
C21H36O2
CAS Number:
Molecular Weight:
320.51

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing