Skip to Content
MilliporeSigma
Chromyl chloride ≥99.99% trace metals basis

Chromyl chloride

Chromyl chloride ≥99.99% trace metals basis
Synonym(s):
Chromium(VI) dichloride dioxide, Chromium(VI) oxychloride, Dichlorochromium(VI) oxide
Linear Formula:
CrO2Cl2
CAS Number:
Molecular Weight:
154.90
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing