Skip to Content
MilliporeSigma
Chromotropic acid disodium salt dihydrate reagent grade, ≥95.0%

Chromotropic acid disodium salt dihydrate

Chromotropic acid disodium salt dihydrate reagent grade, ≥95.0%
Synonym(s):
Chromotropic acid disodium salt dihydrate, 1,8-Dihydroxynaphthalene-3,6-disulfonic acid disodium salt, 4,5-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt
Linear Formula:
(HO)2C10H4(SO3Na)2·2H2O
CAS Number:
Molecular Weight:
400.29

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Chemical Composition
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade