Skip to Content
MilliporeSigma
Chromium(III) nitrate nonahydrate 99%

Chromium(III) nitrate nonahydrate

Chromium(III) nitrate nonahydrate 99%
Synonym(s):
Chromium(III) nitrate nonahydrate
Linear Formula:
Cr(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
400.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-3 of 3
Compare
Product Number
Description
Pricing