Skip to Content
MilliporeSigma
Chromium(III) chloride anhydrous for synthesis

Chromium(III) chloride

Chromium(III) chloride anhydrous for synthesis
Synonym(s):
Chromic chloride, Chromium trichloride, Chromium(III) chloride
Linear Formula:
CrCl3
CAS Number:
Molecular Weight:
158.36

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade