Skip to Content
MilliporeSigma
Chromium(III) acetylacetonate 97%

Chromium(III) acetylacetonate

Chromium(III) acetylacetonate 97%
Synonym(s):
Chromium(III) 2,4-pentanedionate, Cr(acac)3, Chromium(III) acetylacetonate
Linear Formula:
Cr(C5H7O2)3
CAS Number:
Molecular Weight:
349.32

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing