Skip to Content
MilliporeSigma
Chromium(0) hexacarbonyl 98%

Chromium(0) hexacarbonyl

Chromium(0) hexacarbonyl 98%
Synonym(s):
Chromium(0) hexacarbonyl
Linear Formula:
Cr(CO)6
CAS Number:
Molecular Weight:
220.06

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing