Skip to Content
MilliporeSigma
Cholic acid-2,2,4,4-d4 98 atom % D, 98% (CP)

Cholic acid-2,2,4,4-d4

Cholic acid-2,2,4,4-d4 98 atom % D, 98% (CP)
Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholanic acid-D4
Empirical Formula (Hill Notation):
C24D4H36O5
CAS Number:
Molecular Weight:
412.60

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing