Chlortetracycline for system suitability
Synonym(s):
Chlortetracycline hydrochloride, 7-Chlorotetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H23ClN2O8 · HCl
CAS Number:
Molecular Weight:
515.34
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing