Skip to Content
MilliporeSigma
Chlorotriisopropoxytitanium(IV) 95%

Chlorotriisopropoxytitanium(IV)

Chlorotriisopropoxytitanium(IV) 95%
Synonym(s):
Chlorotitanium(IV) triisopropoxide, Titanium(IV) chloride triisopropoxide
Linear Formula:
[(CH3)2CHO]3TiCl
CAS Number:
Molecular Weight:
260.58

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing