Chloroplatinic acid hexahydrate
Synonym(s):
Hexachloroplatinic acid hexahydrate, Hydrogen hexachloroplatinate(IV) hexahydrate, Platinic chloride hexahydrate
Linear Formula:
H2PtCl6 · 6H2O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Reagent Type
Core
Special Grade
Showing 1-3 of 3