Skip to Content
MilliporeSigma
Chlorophyllin sodium copper salt

Chlorophyllin sodium copper salt

Chlorophyllin sodium copper salt
Synonym(s):
Chlorophyllin, coppered trisodium salt
Empirical Formula (Hill Notation):
C34H31CuN4Na3O6
CAS Number:
Molecular Weight:
724.15
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing