Skip to Content
MilliporeSigma
Chlorophenol Red-β-D-galactopyranoside ≥90% (HPLC)

Chlorophenol Red-β-D-galactopyranoside

Chlorophenol Red-β-D-galactopyranoside ≥90% (HPLC)
Synonym(s):
CPRG, Chlorophenol Red-β-D-galactoside
Empirical Formula (Hill Notation):
C25H22Cl2O10S
CAS Number:
Molecular Weight:
585.41

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1