Skip to Content
MilliporeSigma
Chloromethyl pivalate 97%

Chloromethyl pivalate

Chloromethyl pivalate 97%
Synonym(s):
POM-Cl, Pivaloyloxymethyl chloride
Linear Formula:
(CH3)3CCOOCH2Cl
CAS Number:
Molecular Weight:
150.60
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing