Skip to Content
MilliporeSigma
Chloro(dimethyl)thexylsilane 95%

Chloro(dimethyl)thexylsilane

Chloro(dimethyl)thexylsilane 95%
Synonym(s):
Chlorodimethyl(1,1,2-trimethylpropyl)silane, Dimethyl-(2,3-dimethyl-2-butyl)chlorosilane, Dimethyl-thexylchlorosilane, Dimethylthexylsilyl chloride, TDSCl, Thexyldimethylsilyl chloride
Linear Formula:
(CH3)2CHC(CH3)2Si(CH3)2Cl
CAS Number:
Molecular Weight:
178.77

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing