Skip to Content
MilliporeSigma
Chlorodimethylphenethylsilane ≥98%

Chlorodimethylphenethylsilane

Chlorodimethylphenethylsilane ≥98%
Synonym(s):
Chlorodimethyl(2-phenylethyl)silane, Dimethyl(2-phenylethyl)chlorosilane, Dimethyl(phenethyl)chlorosilane
Linear Formula:
C6H5CH2CH2Si(CH3)2Cl
CAS Number:
Molecular Weight:
198.76
EC Number:

Products

Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing