Skip to Content
MilliporeSigma
Chlorhexidine digluconate solution 20% in H2O

Chlorhexidine digluconate solution

Chlorhexidine digluconate solution 20% in H2O
Synonym(s):
1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide)
CAS Number:

Products

Product Category
Brand
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing