Skip to Content
MilliporeSigma
Chloramphenicol British Pharmacopoeia (BP) Reference Standard

Chloramphenicol

Chloramphenicol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloromycetin
Linear Formula:
Cl2CHCONHCH(CH2OH)CH(OH)C6H4NO2
CAS Number:
Molecular Weight:
323.13
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique