Skip to Content
MilliporeSigma
Chiralyst P374 Umicore

Chiralyst P374

Chiralyst P374 Umicore
Synonym(s):
Bis(norbornadiene)rhodium(I) tetrafluoroborate, Rh(nbd)2BF4
Linear Formula:
[Rh(NBD)BF4
CAS Number:
Molecular Weight:
373.99

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing