Skip to Content
MilliporeSigma
Chiralyst P310 Umicore, 99%

Chiralyst P310

Chiralyst P310 Umicore, 99%
Synonym(s):
(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I), Rh(acac)(COD)
Empirical Formula (Hill Notation):
C13H19O2Rh
CAS Number:
Molecular Weight:
310.19

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing