![Cefuroxime sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/111/445/46cf402d-bc5f-4ed1-bd4c-2898cc1698f6/640/46cf402d-bc5f-4ed1-bd4c-2898cc1698f6.png)
Cefuroxime sodium
![Cefuroxime sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/111/445/46cf402d-bc5f-4ed1-bd4c-2898cc1698f6/640/46cf402d-bc5f-4ed1-bd4c-2898cc1698f6.png)
Synonym(s):
Cefuroxime sodium salt
Empirical Formula (Hill Notation):
C16H15N4NaO8S
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing