Skip to Content
MilliporeSigma
Cefotaxime sodium salt potency: 916-964 μg per mg

Cefotaxime sodium salt

Cefotaxime sodium salt potency: 916-964 μg per mg
Synonym(s):
Cefotaxim sodium salt
Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Application
Mode of Action
Technique