Skip to Content
MilliporeSigma
vibrant-m

Cefdinir Related Compound A

vibrant-m
Synonym(s):
(2R)-2-[(Z)-2-(2-Aminothiazol-4-yl)-2-(hydroxyimino)acetamido]-2-[(2RS,5RS)-5-methyl-7-oxo-2,4,5,7-tetrahydro-1H-furo[3,4-d][1,3]thiazin-2-yl]acetic acid
Empirical Formula (Hill Notation):
C14H15N5O6S2
Molecular Weight:
413.43

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1