Skip to Content
MilliporeSigma
Cefadroxil certified reference material, pharmaceutical secondary standard

Cefadroxil

Cefadroxil certified reference material, pharmaceutical secondary standard
Synonym(s):
Cefadroxil monohydrate
Empirical Formula (Hill Notation):
C16H17N3O5S · H2O
CAS Number:
Molecular Weight:
381.40

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade