Skip to Content
MilliporeSigma
CDI reagent grade

CDI

CDI reagent grade
Synonym(s):
1,1′-Carbonyldiimidazole, CDI
Empirical Formula (Hill Notation):
C7H6N4O
CAS Number:
Molecular Weight:
162.15

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing