Skip to Content
MilliporeSigma
Catechol phyproof® Reference Substance

Catechol

Catechol phyproof® Reference Substance
Synonym(s):
Pyrocatechol, 1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol
Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing