Skip to Content
MilliporeSigma
Catalpol phyproof® Reference Substance

Catalpol

Catalpol phyproof® Reference Substance
Synonym(s):
(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside, Catalpinoside, De(p-hydroxybenzoyl)catalposide
Empirical Formula (Hill Notation):
C15H22O10
CAS Number:
Molecular Weight:
362.33
EC Number:

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3