Skip to Content
MilliporeSigma
vibrant-m

Carteolol hydrochloride

vibrant-m
Synonym(s):
Carteolol monohydrochloride
Empirical Formula (Hill Notation):
C16H24N2O3 · HCl
CAS Number:
Molecular Weight:
328.83

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade